Learn More
Biotium Primary Antibody GFAP rGA5 CF680R 1/EA
Catalog No. 5000500463
Encompass_Preferred
GFAP (rGA5) is a recombinant mouse monoclonal antibody that recognizes GFAP This CF680R antibody conjugate has been validated in Immunofluorescence CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability
Catalog No. 50-005-00463 Supplier Biotium Supplier No. P046680R1ML
GFAP (rGA5) is a recombinant mouse monoclonal antibody that recognizes GFAP This CF680R antibody conjugate has been validated in Immunofluorescence CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability