Learn More
Biotium Primary Antibody GFAP rGA5 CF790 1/EA
Catalog No. 5000499766
Encompass_Preferred
GFAP (rGA5) is a recombinant mouse monoclonal antibody that recognizes GFAP This CF790 antibody conjugate has been validated in Immunofluorescence CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability
Catalog No. 50-004-99766 Supplier Biotium Supplier No. P0467901ML
GFAP (rGA5) is a recombinant mouse monoclonal antibody that recognizes GFAP This CF790 antibody conjugate has been validated in Immunofluorescence CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability