Learn More
Biotium Primary Antibody GFAP SdAb2409.GFAP CF583R 1/EA
Catalog No. 5000499360
Encompass_Preferred
GFAP VHH (SdAb2409 GFAP) is a recombinant Alpaca VHH antibody that recognizes single-domain rat GFAP Featuring Biotiums bright and photostable red fluorescent CF583R Dye this antibody conjugate has been validated in Immunofluorescence It is supplied at 200 uL at 0 1 mg/mL
Catalog No. 50-004-99360 Supplier Biotium Supplier No. N001583R200UL
GFAP VHH (SdAb2409 GFAP) is a recombinant Alpaca VHH antibody that recognizes single-domain rat GFAP Featuring Biotiums bright and photostable red fluorescent CF583R Dye this antibody conjugate has been validated in Immunofluorescence It is supplied at 200 uL at 0 1 mg/mL