Learn More
Biotium Primary Antibody GFAP SdAb2409.GFAP CF647 1/EA
Catalog No. 5000499305
Encompass_Preferred
GFAP VHH (SdAb2409 GFAP) is a recombinant Alpaca VHH antibody that recognizes single-domain rat GFAP Featuring Biotiums bright and photostable far-red fluorescent CF647 Dye this antibody conjugate has been validated in Immunofluorescence It is supplied at 200 uL at 0 1 mg/mL
Catalog No. 50-004-99305 Supplier Biotium Supplier No. N001647200UL
GFAP VHH (SdAb2409 GFAP) is a recombinant Alpaca VHH antibody that recognizes single-domain rat GFAP Featuring Biotiums bright and photostable far-red fluorescent CF647 Dye this antibody conjugate has been validated in Immunofluorescence It is supplied at 200 uL at 0 1 mg/mL