Learn More
Biotium Primary Antibody GFAP SdAb2409.GFAP CF680 1/EA
Catalog No. 5000498004
Encompass_Preferred
GFAP VHH (SdAb2409 GFAP) is a recombinant Alpaca VHH antibody that recognizes single-domain rat GFAP Featuring Biotiums bright and photostable far-red fluorescent CF680 Dye this antibody conjugate has been validated in Immunofluorescence It is supplied at 200 uL at 0 1 mg/mL
Catalog No. 50-004-98004 Supplier Biotium Supplier No. N001680200UL
GFAP VHH (SdAb2409 GFAP) is a recombinant Alpaca VHH antibody that recognizes single-domain rat GFAP Featuring Biotiums bright and photostable far-red fluorescent CF680 Dye this antibody conjugate has been validated in Immunofluorescence It is supplied at 200 uL at 0 1 mg/mL