Learn More
Biotium Primary Antibody GFP r9F9.F9 Biotin 1/EA
Catalog No. 5000499971
Encompass_Preferred
GFP (r9F9 F9) is a recombinant mouse monoclonal antibody that recognizes GFP (Green Fluorescent Protein) This biotin antibody conjugate has been validated in Immunofluorescence
Catalog No. 50-004-99971 Supplier Biotium Supplier No. P027BTN100UL
GFP (r9F9 F9) is a recombinant mouse monoclonal antibody that recognizes GFP (Green Fluorescent Protein) This biotin antibody conjugate has been validated in Immunofluorescence