Learn More
Biotium Primary Antibody GFP r9F9.F9 CF740 1/EA
Catalog No. 5000500445
Encompass_Preferred
GFP (r9F9 F9) is a recombinant mouse monoclonal antibody that recognizes GFP (Green Fluorescent Protein) This CF740 antibody conjugate has been validated in Immunofluorescence CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability
Catalog No. 50-005-00445 Supplier Biotium Supplier No. P027740100UL
GFP (r9F9 F9) is a recombinant mouse monoclonal antibody that recognizes GFP (Green Fluorescent Protein) This CF740 antibody conjugate has been validated in Immunofluorescence CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability