Learn More
Biotium Primary Antibody GFP r9F9.F9 CF790 1/EA
Catalog No. 5000500782
Encompass_Preferred
GFP (r9F9 F9) is a recombinant mouse monoclonal antibody that recognizes GFP (Green Fluorescent Protein) This CF790 antibody conjugate has been validated in Immunofluorescence CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability
Catalog No. 50-005-00782 Supplier Biotium Supplier No. P027790100UL
GFP (r9F9 F9) is a recombinant mouse monoclonal antibody that recognizes GFP (Green Fluorescent Protein) This CF790 antibody conjugate has been validated in Immunofluorescence CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability