Learn More
Biotium Primary Antibody GFP r9F9.F9 Purified BSA free 5 mg mL 1/EA
Supplier:  Biotium P0275MG
Encompass_Preferred
GFP (r9F9 F9) is a recombinant mouse monoclonal antibody that recognizes GFP (Green Fluorescent Protein) This purified antibody has been validated in Immunofluorescence It is supplied at 5 mg/mL BSA-free and azide-free
Catalog No. 50-005-00378