Learn More
Biotium Primary Antibody Human IgG Fc N2508.HFC CF488A 1/EA
Catalog No. 5000498499
Encompass_Preferred
Anti-Human IgG (Fc) rVHH (N2508 HFC) is a recombinant Alpaca VHH antibody that recognizes the Fc domain of human IgG and is validated for IF and flow Green fluorescent STORM-compatible CF488A dye has Ex/Em at 490/515 nm Unit size 500 uL at 0 1 mg/mL
Catalog No. 50-004-98499 Supplier Biotium Supplier No. N007488A500UL
Anti-Human IgG (Fc) rVHH (N2508 HFC) is a recombinant Alpaca VHH antibody that recognizes the Fc domain of human IgG and is validated for IF and flow Green fluorescent STORM-compatible CF488A dye has Ex/Em at 490/515 nm Unit size 500 uL at 0 1 mg/mL