Learn More
Biotium Primary Antibody Human IgG Fc N2508.HFC CF568 1/EA
Catalog No. 5000498576
Encompass_Preferred
Anti-Human IgG (Fc) rVHH (N2508 HFC) is a recombinant Alpaca VHH antibody that recognizes the Fc domain of human IgG and is validated for IF and flow Red fluorescent STORM-compatible CF568 dye has Ex/Em at 562/583 nm Unit size 500 uL at 0 1 mg/mL
Catalog No. 50-004-98576 Supplier Biotium Supplier No. N007568500UL
Anti-Human IgG (Fc) rVHH (N2508 HFC) is a recombinant Alpaca VHH antibody that recognizes the Fc domain of human IgG and is validated for IF and flow Red fluorescent STORM-compatible CF568 dye has Ex/Em at 562/583 nm Unit size 500 uL at 0 1 mg/mL