Learn More
Biotium Primary Antibody Human IgG Fc N2508.HFC CF594 1/EA
Catalog No. 5000498504
Encompass_Preferred
Anti-Human IgG (Fc) rVHH (N2508 HFC) is a recombinant Alpaca VHH antibody that recognizes the Fc domain of human IgG and is validated for IF and flow Red fluorescent photostable CF594 dye has Ex/Em at 593/614 nm Unit size 500 uL at 0 1 mg/mL
Catalog No. 50-004-98504 Supplier Biotium Supplier No. N007594500UL
Anti-Human IgG (Fc) rVHH (N2508 HFC) is a recombinant Alpaca VHH antibody that recognizes the Fc domain of human IgG and is validated for IF and flow Red fluorescent photostable CF594 dye has Ex/Em at 593/614 nm Unit size 500 uL at 0 1 mg/mL