Learn More
Biotium Primary Antibody Human IgG Fc N2508.HFC CF640R 1/EA
Catalog No. 5000497489
Encompass_Preferred
Anti-Human IgG (Fc) rVHH (N2508 HFC) is a recombinant Alpaca VHH antibody that recognizes the Fc domain of human IgG and is validated for IF and flow Far-red fluorescent photostable CF640R dye has Ex/Em at 642/662 nm Unit size 500 uL at 0 1 mg/mL
Catalog No. 50-004-97489 Supplier Biotium Supplier No. N007640R500UL
Anti-Human IgG (Fc) rVHH (N2508 HFC) is a recombinant Alpaca VHH antibody that recognizes the Fc domain of human IgG and is validated for IF and flow Far-red fluorescent photostable CF640R dye has Ex/Em at 642/662 nm Unit size 500 uL at 0 1 mg/mL