Learn More
Biotium Primary Antibody Human IgG Fc N2508.HFC CF680 1/EA
Catalog No. 5000498512
Encompass_Preferred
Anti-Human IgG (Fc) rVHH (N2508 HFC) is a recombinant Alpaca VHH antibody that recognizes the Fc domain of human IgG and is validated for IF and flow Far-red/near-IR STORM-compatible CF680 dye has Ex/Em at 681/698 nm Unit size 500 uL at 0 1 mg/mL
Catalog No. 50-004-98512 Supplier Biotium Supplier No. N007680500UL
Anti-Human IgG (Fc) rVHH (N2508 HFC) is a recombinant Alpaca VHH antibody that recognizes the Fc domain of human IgG and is validated for IF and flow Far-red/near-IR STORM-compatible CF680 dye has Ex/Em at 681/698 nm Unit size 500 uL at 0 1 mg/mL