Learn More
Biotium Primary Antibody Human IgG Fc N2508.HFC CF740 1/EA
Catalog No. 5000498513
Encompass_Preferred
Anti-Human IgG (Fc) rVHH (N2508 HFC) is a recombinant Alpaca VHH antibody that recognizes the Fc domain of human IgG and is validated for IF and flow Near-IR photostable CF740 dye has Ex/Em at 742/767 nm Unit size 500 uL at 0 1 mg/mL
Catalog No. 50-004-98513 Supplier Biotium Supplier No. N007740500UL
Anti-Human IgG (Fc) rVHH (N2508 HFC) is a recombinant Alpaca VHH antibody that recognizes the Fc domain of human IgG and is validated for IF and flow Near-IR photostable CF740 dye has Ex/Em at 742/767 nm Unit size 500 uL at 0 1 mg/mL