Learn More
Biotium Methicillin Resistant Staphylococcus Aureus (MRSA) (332/423), CF740 conjugate, 0.1mg/mL
Supplier:  Biotium BNC740229100
Encompass_Preferred
Staphylococcal enterotoxins represent a group of proteins, which are secreted by Staphylococcus aureus and cause the intoxication staphylococcal food poisoning syndrome. The illness characterized by high fever, hypotension, diarrhea, shock, and in some cases death. Their molecular masses range between 27 and 30 kDa. At present, seven enterotoxins are known, namely A, B, C1, C2, C3, D and E. Their amino acid sequences have been determined and it was shown that all are single chain polypeptides containing one disulfide bond formed by two half cysteine located in the middle of the polypeptide chain, which form the so called cysteine loop. Enterotoxins are extremely potent activator of T cells, stimulating the production and secretion of various cytokines, which mediate many of the toxic effects of these substances. Enterotoxins are super antigens, inducing polyclonal T cell activation by binding to the TCR and to the alpha chain of the MHC II molecule simultaneously.Primary an
Catalog No. 50-005-00578