Learn More
Biotium Primary Antibody Mitochondrial Marker 2509.MTC CF488A 1/EA
Catalog No. 5000499742
Encompass_Preferred
Mitochondrial Marker (2509 MTC) is a recombinant mouse monoclonal antibody that recognizes a 60 kDa antigen associated with the mitochondria in cells This CF488A antibody conjugate has been validated in IF CF and reg dyes are fluorescent dyes with advantages in brightness and photostability
Catalog No. 50-004-99742 Supplier Biotium Supplier No. P045488A500UL
Mitochondrial Marker (2509 MTC) is a recombinant mouse monoclonal antibody that recognizes a 60 kDa antigen associated with the mitochondria in cells This CF488A antibody conjugate has been validated in IF CF and reg dyes are fluorescent dyes with advantages in brightness and photostability