Learn More
Biotium Primary Antibody Mitochondrial Marker 2509.MTC CF640R 1/EA
Catalog No. 5000499999
Encompass_Preferred
Mitochondrial Marker (2509 MTC) is a recombinant mouse monoclonal antibody that recognizes a 60 kDa antigen associated with the mitochondria in cells This CF640R antibody conjugate has been validated in IF CF and reg dyes are fluorescent dyes with advantages in brightness and photostability
Catalog No. 50-004-99999 Supplier Biotium Supplier No. P045640R500UL
Mitochondrial Marker (2509 MTC) is a recombinant mouse monoclonal antibody that recognizes a 60 kDa antigen associated with the mitochondria in cells This CF640R antibody conjugate has been validated in IF CF and reg dyes are fluorescent dyes with advantages in brightness and photostability