Learn More
Biotium Primary Antibody Mitochondrial Marker 2509.MTC CF647 1/EA
Catalog No. 5000501321
Encompass_Preferred
Mitochondrial Marker (2509 MTC) is a recombinant mouse monoclonal antibody that recognizes a 60 kDa antigen associated with the mitochondria in cells This CF647 antibody conjugate has been validated in IF CF and reg dyes are fluorescent dyes with advantages in brightness and photostability
Catalog No. 50-005-01321 Supplier Biotium Supplier No. P045647500UL
Mitochondrial Marker (2509 MTC) is a recombinant mouse monoclonal antibody that recognizes a 60 kDa antigen associated with the mitochondria in cells This CF647 antibody conjugate has been validated in IF CF and reg dyes are fluorescent dyes with advantages in brightness and photostability