Learn More
Biotium Primary Antibody Mitochondrial Marker 2509.MTC CF680R 1/EA
Catalog No. 5000500000
Encompass_Preferred
Mitochondrial Marker (2509 MTC) is a recombinant mouse monoclonal antibody that recognizes a 60 kDa antigen associated with the mitochondria in cells This CF680R antibody conjugate has been validated in IF CF and reg dyes are fluorescent dyes with advantages in brightness and photostability
Catalog No. 50-005-00000 Supplier Biotium Supplier No. P045680R500UL
Mitochondrial Marker (2509 MTC) is a recombinant mouse monoclonal antibody that recognizes a 60 kDa antigen associated with the mitochondria in cells This CF680R antibody conjugate has been validated in IF CF and reg dyes are fluorescent dyes with advantages in brightness and photostability