Learn More
Biotium Primary Antibody Mitochondrial Marker 2509.MTC CF790 1/EA
Catalog No. 5000500713
Encompass_Preferred
Mitochondrial Marker (2509 MTC) is a recombinant mouse monoclonal antibody that recognizes a 60 kDa antigen associated with the mitochondria in cells This CF790 antibody conjugate has been validated in IF CF and reg dyes are fluorescent dyes with advantages in brightness and photostability
Catalog No. 50-005-00713 Supplier Biotium Supplier No. P045790500UL
Mitochondrial Marker (2509 MTC) is a recombinant mouse monoclonal antibody that recognizes a 60 kDa antigen associated with the mitochondria in cells This CF790 antibody conjugate has been validated in IF CF and reg dyes are fluorescent dyes with advantages in brightness and photostability