Learn More
Biotium Primary Antibody Mouse Fab kappa N2507.MFAB CF488A 1/EA
Catalog No. 5000498449
Encompass_Preferred
Alpaca Anti-Mouse (Fab) Kappa rVHH (N2507 MFAB) is a recombinant Alpaca VHH antibody that recognizes the mouse kappa light chain and is validated for IF flow and WB Green fluorescent STORM-compatible CF488A dye has Ex/Em at 488A/519 nm Unit size 500 uL at 0 1 mg/mL
Catalog No. 50-004-98449 Supplier Biotium Supplier No. N005488A500UL
Alpaca Anti-Mouse (Fab) Kappa rVHH (N2507 MFAB) is a recombinant Alpaca VHH antibody that recognizes the mouse kappa light chain and is validated for IF flow and WB Green fluorescent STORM-compatible CF488A dye has Ex/Em at 488A/519 nm Unit size 500 uL at 0 1 mg/mL