Learn More
Biotium Primary Antibody Mouse Fab kappa N2507.MFAB CF568 1/EA
Supplier:  Biotium N005568500UL
Encompass_Preferred
Alpaca Anti-Mouse (Fab) Kappa rVHH (N2507 MFAB) is a recombinant Alpaca VHH antibody that recognizes the mouse kappa light chain and is validated for IF flow and WB Red fluorescent STORM-compatible CF568 dye has Ex/Em at 562/583 nm Unit size 500 uL at 0 1 mg/mL
Catalog No. 50-004-98595