Learn More
Biotium Primary Antibody Mouse IgG1 Fc N2504.M1FC CF594 1/EA
Catalog No. 5000499309
Encompass_Preferred
Anti-Mouse IgG1 (Fc) rVHH (N2504 M1FC) is a recombinant Alpaca VHH antibody that recognizes the Fc fragment of mouse IgG1 and is validated for IF flow and WB Red fluorescent photostable CF594 dye has Ex/Em at 593/614 nm Unit size 500 uL at 0 1 mg/mL
Catalog No. 50-004-99309 Supplier Biotium Supplier No. N004594500UL
Anti-Mouse IgG1 (Fc) rVHH (N2504 M1FC) is a recombinant Alpaca VHH antibody that recognizes the Fc fragment of mouse IgG1 and is validated for IF flow and WB Red fluorescent photostable CF594 dye has Ex/Em at 593/614 nm Unit size 500 uL at 0 1 mg/mL