Learn More
Biotium Primary Antibody Mouse IgG1 Fc N2504.M1FC CF680 1/EA
Catalog No. 5000497498
Encompass_Preferred
Anti-Mouse IgG1 (Fc) rVHH (N2504 M1FC) is a recombinant Alpaca VHH antibody that recognizes the Fc fragment of mouse IgG1 and is validated for IF flow and WB Far-red/near-IR STORM-compatible CF680 dye has Ex/Em at 681/698 nm Unit size 500 uL at 0 1 mg/mL
Catalog No. 50-004-97498 Supplier Biotium Supplier No. N004680500UL
Anti-Mouse IgG1 (Fc) rVHH (N2504 M1FC) is a recombinant Alpaca VHH antibody that recognizes the Fc fragment of mouse IgG1 and is validated for IF flow and WB Far-red/near-IR STORM-compatible CF680 dye has Ex/Em at 681/698 nm Unit size 500 uL at 0 1 mg/mL