Learn More
Biotium Primary Antibody NeuN rA60 CF640R 1/EA
Catalog No. 5000500805
Encompass_Preferred
NeuN (rA60) is a recombinant mouse monoclonal antibody that recognizes neuron-specific protein NeuN This CF640R antibody conjugate has been validated in Immunofluorescence CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability
Catalog No. 50-005-00805 Supplier Biotium Supplier No. P037640R500UL
Add to Cart
Add to Cart
NeuN (rA60) is a recombinant mouse monoclonal antibody that recognizes neuron-specific protein NeuN This CF640R antibody conjugate has been validated in Immunofluorescence CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability