Learn More
Biotium Primary Antibody NeuN rA60 CF740 1/EA
Catalog No. 5000500806
Encompass_Preferred
NeuN (rA60) is a recombinant mouse monoclonal antibody that recognizes neuron-specific protein NeuN This CF740 antibody conjugate has been validated in Immunofluorescence CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability
Catalog No. 50-005-00806 Supplier Biotium Supplier No. P037740500UL
NeuN (rA60) is a recombinant mouse monoclonal antibody that recognizes neuron-specific protein NeuN This CF740 antibody conjugate has been validated in Immunofluorescence CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability