Learn More
Biotium Primary Antibody Neurofilament H rRmdO-20 CF680R 1/EA
Catalog No. 5000500799
Encompass_Preferred
Neurofilament-H (rRmdO-20) is a recombinant mouse monoclonal antibody that recognizes Neurofilament-H This CF680R antibody conjugate has been validated in Immunofluorescence CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability
Catalog No. 50-005-00799 Supplier Biotium Supplier No. P035680R500UL
Neurofilament-H (rRmdO-20) is a recombinant mouse monoclonal antibody that recognizes Neurofilament-H This CF680R antibody conjugate has been validated in Immunofluorescence CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability