Learn More
Biotium Primary Antibody Nuclear membrane 2406.NM CF488A 1/EA
Catalog No. 5000497528
Encompass_Preferred
This recombinant monoclonal mouse antibody conjugate stains the nuclear membrane in human cells Featuring Biotiums bright and photostable green fluorescent CF488A Dye (Ex/Em 490/515 nm) 100 ug/mL 1 mL
Catalog No. 50-004-97528 Supplier Biotium Supplier No. P023488A1ML
This recombinant monoclonal mouse antibody conjugate stains the nuclear membrane in human cells Featuring Biotiums bright and photostable green fluorescent CF488A Dye (Ex/Em 490/515 nm) 100 ug/mL 1 mL