Learn More
Biotium Primary Antibody Nuclear membrane 2406.NM CF568 1/EA
Catalog No. 5000497188
Encompass_Preferred
This recombinant monoclonal mouse antibody conjugate stains the nuclear membrane in human cells Featuring Biotiums bright and photostable red fluorescent CF568 Dye (Ex/Em 562/583 nm) 100 ug/mL 1 mL
Catalog No. 50-004-97188 Supplier Biotium Supplier No. P0235681ML
This recombinant monoclonal mouse antibody conjugate stains the nuclear membrane in human cells Featuring Biotiums bright and photostable red fluorescent CF568 Dye (Ex/Em 562/583 nm) 100 ug/mL 1 mL