Learn More
Biotium Primary Antibody Nuclear membrane 2406.NM CF568 1/EA
Catalog No. 5000499111
Encompass_Preferred
This recombinant monoclonal mouse antibody conjugate stains the nuclear membrane in human cells Featuring Biotiums bright and photostable red fluorescent CF568 Dye (Ex/Em 562/583 nm) 100 ug/mL 200 uL
Catalog No. 50-004-99111 Supplier Biotium Supplier No. P023568200UL
This recombinant monoclonal mouse antibody conjugate stains the nuclear membrane in human cells Featuring Biotiums bright and photostable red fluorescent CF568 Dye (Ex/Em 562/583 nm) 100 ug/mL 200 uL