Learn More
Biotium Primary Antibody Nuclear membrane 2406.NM CF594 1/EA
Catalog No. 5000499049
Encompass_Preferred
This recombinant monoclonal mouse antibody conjugate stains the nuclear membrane in human cells Featuring Biotiums bright and photostable deep red fluorescent CF594 Dye (Ex/Em 593/614 nm) 100 ug/mL 1 mL
Catalog No. 50-004-99049 Supplier Biotium Supplier No. P0235941ML
This recombinant monoclonal mouse antibody conjugate stains the nuclear membrane in human cells Featuring Biotiums bright and photostable deep red fluorescent CF594 Dye (Ex/Em 593/614 nm) 100 ug/mL 1 mL