Learn More
Biotium Primary Antibody Nucleolin 2507.NCL CF647 1/EA
Catalog No. 5000499484
Encompass_Preferred
Nucleolin (2507 NCL) is a recombinant mouse monoclonal antibody that recognizes nucleolin This CF647 antibody conjugate has been validated in Immunofluorescence CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability
Catalog No. 50-004-99484 Supplier Biotium Supplier No. P039647500UL
Nucleolin (2507 NCL) is a recombinant mouse monoclonal antibody that recognizes nucleolin This CF647 antibody conjugate has been validated in Immunofluorescence CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability