Learn More
Biotium Primary Antibody Podocalyxin 3D3 CF740 1/EA
Supplier:  Biotium BNC740427500
Encompass_Preferred
Podocalyxin is a member of the CD34 transmembrane sialomucin family It is highly on kidney podocyte foot projections and plays essential roles in kidney development and homeostasis blood filtration and urine formation It is also expressed on vascular endothelia hematopoietic progenitors and a subset of neurons Overexpression of podocalyxin may be linked to more aggressive tumor behavior Podocalyxin antibody can identify podocytes in the urine (podocyturia) that may indicate glomerular disease pre-eclampsia and other kidney pathology Primary antibodies are available purified or with a selection of fluorescent CF Dyes and other labels CF Dyes offer exceptional brightness and photostability Note Conjugates of blue fluorescent dyes like CF405S and CF405M are not recommended for detecting low abundance targets because blue dyes have lower fluorescence and can give higher non-specific background than other dye colors
Catalog No. 50-004-96931