Learn More
Biotium Primary Antibody PSD95 rK28/43 CF640R 1/EA
Catalog No. 5000499684
Encompass_Preferred
PSD95 (rK28/43)is a recombinant mouse monoclonal antibody that recognizes PSD95 This CF640R antibody conjugate has been validated in Immunofluorescence CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability
Catalog No. 50-004-99684 Supplier Biotium Supplier No. P036640R500UL
PSD95 (rK28/43)is a recombinant mouse monoclonal antibody that recognizes PSD95 This CF640R antibody conjugate has been validated in Immunofluorescence CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability