Learn More
Biotium Primary Antibody PSD95 rK28/43 CF647 1/EA
Catalog No. 5000499467
Encompass_Preferred
PSD95 (rK28/43)is a recombinant mouse monoclonal antibody that recognizes PSD95 This CF647 antibody conjugate has been validated in Immunofluorescence CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability
Catalog No. 50-004-99467 Supplier Biotium Supplier No. P036647500UL
PSD95 (rK28/43)is a recombinant mouse monoclonal antibody that recognizes PSD95 This CF647 antibody conjugate has been validated in Immunofluorescence CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability