Learn More
Biotium Primary Antibody PSD95 rK28/43 CF680R 1/EA
Catalog No. 5000500383
Encompass_Preferred
PSD95 (rK28/43)is a recombinant mouse monoclonal antibody that recognizes PSD95 This CF680R antibody conjugate has been validated in Immunofluorescence CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability
Catalog No. 50-005-00383 Supplier Biotium Supplier No. P036680R500UL
PSD95 (rK28/43)is a recombinant mouse monoclonal antibody that recognizes PSD95 This CF680R antibody conjugate has been validated in Immunofluorescence CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability