Learn More
Biotium Primary Antibody Rabbit IgG Fc N2507.RFC CF594 1/EA
Catalog No. 5000499306
Encompass_Preferred
Alpaca Anti-Rabbit IgG (Fc) rVHH (N2507 RFC) is a recombinant Alpaca VHH antibody that recognizes the Fc fragment of rabbit IgG and is validated for immunofluorescence Red fluorescent photostable CF594 dye has Ex/Em at 593/614 nm Unit size 500 uL at 0 1 mg/mL
Catalog No. 50-004-99306 Supplier Biotium Supplier No. N006594500UL
Alpaca Anti-Rabbit IgG (Fc) rVHH (N2507 RFC) is a recombinant Alpaca VHH antibody that recognizes the Fc fragment of rabbit IgG and is validated for immunofluorescence Red fluorescent photostable CF594 dye has Ex/Em at 593/614 nm Unit size 500 uL at 0 1 mg/mL