Learn More
Biotium Primary Antibody Rabbit IgG Fc N2507.RFC CF640R 1/EA
Catalog No. 5000499363
Encompass_Preferred
Alpaca Anti-Rabbit IgG (Fc) rVHH (N2507 RFC) is a recombinant Alpaca VHH antibody that recognizes the Fc fragment of rabbit IgG and is validated for immunofluorescence Far-red fluorescent photostable CF640R dye has Ex/Em at 642/662 nm Unit size 500 uL at 0 1 mg/mL
Catalog No. 50-004-99363 Supplier Biotium Supplier No. N006640R500UL
Add to Cart
Add to Cart
Alpaca Anti-Rabbit IgG (Fc) rVHH (N2507 RFC) is a recombinant Alpaca VHH antibody that recognizes the Fc fragment of rabbit IgG and is validated for immunofluorescence Far-red fluorescent photostable CF640R dye has Ex/Em at 642/662 nm Unit size 500 uL at 0 1 mg/mL