Learn More
Biotium Primary Antibody Rabbit IgG Fc N2507.RFC CF680 1/EA
Catalog No. 5000498199
Encompass_Preferred
Alpaca Anti-Rabbit IgG (Fc) rVHH (N2507 RFC) is a recombinant Alpaca VHH antibody that recognizes the Fc fragment of rabbit IgG and is validated for immunofluorescence Far-red/near-IR STORM-compatible CF680 dye has Ex/Em at 681/698 nm Unit size 500 uL at 0 1 mg/mL
Catalog No. 50-004-98199 Supplier Biotium Supplier No. N006680500UL
Alpaca Anti-Rabbit IgG (Fc) rVHH (N2507 RFC) is a recombinant Alpaca VHH antibody that recognizes the Fc fragment of rabbit IgG and is validated for immunofluorescence Far-red/near-IR STORM-compatible CF680 dye has Ex/Em at 681/698 nm Unit size 500 uL at 0 1 mg/mL