Learn More
Biotium Primary Antibody Rabbit IgG Fc N2507.RFC CF680R 1/EA
Catalog No. 5000499310
Encompass_Preferred
Alpaca Anti-Rabbit IgG (Fc) rVHH (N2507 RFC) is a recombinant Alpaca VHH antibody that recognizes the Fc fragment of rabbit IgG and is validated for immunofluorescence Far-red/near-IR photostable CF680R dye has Ex/Em at 680/701 nm Unit size 500 uL at 0 1 mg/mL
Catalog No. 50-004-99310 Supplier Biotium Supplier No. N006680R500UL
Alpaca Anti-Rabbit IgG (Fc) rVHH (N2507 RFC) is a recombinant Alpaca VHH antibody that recognizes the Fc fragment of rabbit IgG and is validated for immunofluorescence Far-red/near-IR photostable CF680R dye has Ex/Em at 680/701 nm Unit size 500 uL at 0 1 mg/mL