Learn More
Biotium Primary Antibody SYT1 cytoplasmic domain SdAb2501.SYT1 CF647 1/EA
Catalog No. 5000498346
Encompass_Preferred
SYT1 VHH (SdAb2501 SYT1) is a recombinant Alpaca VHH antibody that recognizes single-domain rat and mouse SYT1 Featuring Biotiums bright and photostable far-red fluorescent CF647 Dye this antibody conjugate has been validated in Immunofluorescence It is supplied at 200 uL at 0 1 mg/mL
Catalog No. 50-004-98346 Supplier Biotium Supplier No. N002647200UL
SYT1 VHH (SdAb2501 SYT1) is a recombinant Alpaca VHH antibody that recognizes single-domain rat and mouse SYT1 Featuring Biotiums bright and photostable far-red fluorescent CF647 Dye this antibody conjugate has been validated in Immunofluorescence It is supplied at 200 uL at 0 1 mg/mL