Learn More
Biotium Primary Antibody VGLUT1 SdAb2412.VGLUT1 CF647 1/EA
Catalog No. 5000498201
Encompass_Preferred
VGLUT1 VHH (SdAb2412 VGLUT1) is a recombinant Alpaca VHH antibody that recognizes single-domain rat and mouse VGLUT1 Featuring Biotiums bright and photostable far-red fluorescent CF647 Dye this antibody conjugate has been validated in Immunofluorescence It is supplied at 200 uL at 0 1 mg/mL
Catalog No. 50-004-98201 Supplier Biotium Supplier No. N003647200UL
VGLUT1 VHH (SdAb2412 VGLUT1) is a recombinant Alpaca VHH antibody that recognizes single-domain rat and mouse VGLUT1 Featuring Biotiums bright and photostable far-red fluorescent CF647 Dye this antibody conjugate has been validated in Immunofluorescence It is supplied at 200 uL at 0 1 mg/mL