Learn More
Biotium Primary Antibody Vimentin rV9 CF640R 1/EA
Catalog No. 5000499743
Encompass_Preferred
Vimentin (rV9) is a recombinant mouse monoclonal antibody that recognizes Vimentin This CF640R antibody conjugate has been validated in Immunofluorescence CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability
Catalog No. 50-004-99743 Supplier Biotium Supplier No. P044640R500UL
Vimentin (rV9) is a recombinant mouse monoclonal antibody that recognizes Vimentin This CF640R antibody conjugate has been validated in Immunofluorescence CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability