Learn More
Biotium Primary Antibody ZO 1 rZO1-1A12 CF594 1/EA
Catalog No. 5000500707
Encompass_Preferred
ZO-1 (rZO1-1A12) is a recombinant mouse monoclonal antibody that recognizes ZO-1 This CF594 antibody conjugate has been validated in Immunofluorescence CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability
Catalog No. 50-005-00707 Supplier Biotium Supplier No. P034594500UL
Add to Cart
Add to Cart
ZO-1 (rZO1-1A12) is a recombinant mouse monoclonal antibody that recognizes ZO-1 This CF594 antibody conjugate has been validated in Immunofluorescence CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability