Learn More
ATCC Primary Aortic Endothelial Cells; Normal, Human (Homo sapiens)
Supplier:  ATCC PCS100011
Encompass
Normal Primary Human Aortic Endothelial Cells, when grown in Vascular Cell Basal Media supplemented with Endothelial Cell Growth Kit components, provide an ideal cell system to propagate aortic endothelial cells in low serum conditions with or without the addition of human recombinant VEGF.
Catalog No. 50-238-4734