Learn More
ATCC Primary Aortic Smooth Muscle Cells; Normal, Human
Catalog No. 502384740
Encompass
Normal Primary Aortic Smooth Muscle Cells, when grown in Vascular Cell Basal Medium supplemented with Vascular Smooth Muscle Growth Kit components, provide an ideal cell system to propagate aortic smooth muscle cells in low serum conditions.
Catalog No. 50-238-4740 Supplier ATCC Supplier No. PCS100012
Normal Primary Aortic Smooth Muscle Cells, when grown in Vascular Cell Basal Medium supplemented with Vascular Smooth Muscle Growth Kit components, provide an ideal cell system to propagate aortic smooth muscle cells in low serum conditions.