Learn More
ATCC Primary Broncheal/Tracheal Smooth Muscle Cells; Normal, Human
Catalog No. 502382273
Encompass
Primary Bronchial Tracheal Smooth Muscle Cells are myocytes isolated from the respiratory tract of a donor and can be used in the study of angiogenesis, atherosclerosis, diabetes, or vascular/pulmonary biology.
Catalog No. 50-238-2273 Supplier ATCC Supplier No. PCS130011
 Restricted Item
Add to Cart
Add to Cart
Primary Bronchial Tracheal Smooth Muscle Cells are myocytes isolated from the respiratory tract of a donor and can be used in the study of angiogenesis, atherosclerosis, diabetes, or vascular/pulmonary biology.