Learn More
ATCC Primary Bronchial/Tracheal Epithelial Cells; COPD, Human
Catalog No. 502381882
Encompass
Primary bronchial/tracheal epithelial cells COPD are spindle-shaped cells isolated from the respiratory tract of a donor with COPD. The cells have an application in microbial (eg, viral, bacterial) infection and pathogenesis, airway inflammation and wound healing, asthma, pulmonary fibrosis, chronic obstructive pulmonary disease, chronic bronchitis, toxicology, and testing of pharmaceuticals.
Catalog No. 50-238-1882 Supplier ATCC Supplier No. PCS300013
Primary bronchial/tracheal epithelial cells COPD are spindle-shaped cells isolated from the respiratory tract of a donor with COPD. The cells have an application in microbial (eg, viral, bacterial) infection and pathogenesis, airway inflammation and wound healing, asthma, pulmonary fibrosis, chronic obstructive pulmonary disease, chronic bronchitis, toxicology, and testing of pharmaceuticals.