Learn More
ATCC Primary Bronchial/Tracheal Epithelial Cells; Normal, Human
Catalog No. 502382292
Encompass
Primary Bronchial/Tracheal Epithelial Cells; Normal, Human, when grown in Airway Epithelial Cell Basal Media supplemented with Bronchial/Tracheal Epithelial Cell Growth Kit components, provide an ideal cell system to propagate bronchial/tracheal epithelial cells in serum-free conditions.
Catalog No. 50-238-2292 Supplier ATCC Supplier No. PCS300010
Primary Bronchial/Tracheal Epithelial Cells; Normal, Human, when grown in Airway Epithelial Cell Basal Media supplemented with Bronchial/Tracheal Epithelial Cell Growth Kit components, provide an ideal cell system to propagate bronchial/tracheal epithelial cells in serum-free conditions.